RIE156328244 Rieke Metals, Inc.

3',5'-dimethyl-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843789
  • Cas Number : 898754-92-2
  • Molecular Weight : 266.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-31
Availability details for
Quantity
  • Availability
  • Add to Cart