RIE156328247 Rieke Metals, Inc.

3'-chloro-3-(2,6-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843792
  • Cas Number : 898754-98-8
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-(2,6-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-4-3-5-12(2)14(11)7-9-17(20)13-6-8-16(19)15(18)10-13/h3-6,8,10H,7,9H2,1-2H3
Inchi Key SUXBVVOLIJJRHR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-34
Availability details for
Quantity
  • Availability
  • Add to Cart