RIE156328257 Rieke Metals, Inc.

2',3'-dichloro-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843802
  • Cas Number : 898755-18-5
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,3-dichlorophenyl)-3-(2,6-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16Cl2O/c1-11-5-3-6-12(2)13(11)9-10-16(20)14-7-4-8-15(18)17(14)19/h3-8H,9-10H2,1-2H3
Inchi Key PTTBKMCUJRHHON-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-53
Availability details for
Quantity
  • Availability
  • Add to Cart