RIE156328261 Rieke Metals, Inc.

3',5'-dichloro-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843806
  • Cas Number : 898755-26-5
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dichlorophenyl)-3-(2,6-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16Cl2O/c1-11-4-3-5-12(2)16(11)6-7-17(20)13-8-14(18)10-15(19)9-13/h3-5,8-10H,6-7H2,1-2H3
Inchi Key JJHHSWQLAHFQEC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-57
Availability details for
Quantity
  • Availability
  • Add to Cart