RIE156328262 Rieke Metals, Inc.

2',4'-difluoro-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843807
  • Cas Number : 898755-28-7
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-58
Availability details for
Quantity
  • Availability
  • Add to Cart