RIE156328263 Rieke Metals, Inc.

3',4'-difluoro-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843808
  • Cas Number : 898755-30-1
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-59
Availability details for
Quantity
  • Availability
  • Add to Cart