RIE156328265 Rieke Metals, Inc.

3-(2,6-dimethylphenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843810
  • Cas Number : 898755-34-5
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-61
Availability details for
Quantity
  • Availability
  • Add to Cart