RIE156328268 Rieke Metals, Inc.

cyclobutyl 2-(2,6-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843812
  • Cas Number : 898755-43-6
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2902-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2902-00
Availability details for
Quantity
  • Availability
  • Add to Cart