RIE156328283 Rieke Metals, Inc.

4'-carboethoxy-3-(3,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843827
  • Cas Number : 898779-05-0
  • Molecular Weight : 310.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[3-(3,4-dimethylphenyl)propanoyl]benzoate
Inchi Code 1S/C20H22O3/c1-4-23-20(22)18-10-8-17(9-11-18)19(21)12-7-16-6-5-14(2)15(3)13-16/h5-6,8-11,13H,4,7,12H2,1-3H3
Inchi Key CSQVBTMPNMQNLA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-13
Availability details for
Quantity
  • Availability
  • Add to Cart