RIE156328284 Rieke Metals, Inc.

3-(3,4-dimethylphenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD03843828
  • Cas Number : 898779-08-3
  • Molecular Weight : 284.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3,4-dimethylphenyl)-1-[2-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C18H20OS/c1-13-8-9-15(12-14(13)2)10-11-17(19)16-6-4-5-7-18(16)20-3/h4-9,12H,10-11H2,1-3H3
Inchi Key VWDLQGCQIRZNHS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-14
Availability details for
Quantity
  • Availability
  • Add to Cart