RIE156328290 Rieke Metals, Inc.

3-(3,4-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843834
  • Cas Number : 898779-26-5
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-22
Availability details for
Quantity
  • Availability
  • Add to Cart