RIE156328291 Rieke Metals, Inc.

3-(3,4-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843835
  • Cas Number : 898779-29-8
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-23
Availability details for
Quantity
  • Availability
  • Add to Cart