RIE156328295 Rieke Metals, Inc.

2',6'-dimethyl-3-(3,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843839
  • Cas Number : 898779-41-4
  • Molecular Weight : 266.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-dimethylphenyl)-3-(3,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C19H22O/c1-13-8-9-17(12-16(13)4)10-11-18(20)19-14(2)6-5-7-15(19)3/h5-9,12H,10-11H2,1-4H3
Inchi Key FBSVZCXRYBRWSO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-29
Availability details for
Quantity
  • Availability
  • Add to Cart