RIE156328297 Rieke Metals, Inc.

3',5'-dimethyl-3-(3,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843841
  • Cas Number : 898779-47-0
  • Molecular Weight : 266.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3,4-dimethylphenyl)-1-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C19H22O/c1-13-9-14(2)11-18(10-13)19(20)8-7-17-6-5-15(3)16(4)12-17/h5-6,9-12H,7-8H2,1-4H3
Inchi Key AXWVCMCYTDTOBX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-31
Availability details for
Quantity
  • Availability
  • Add to Cart