RIE156328298 Rieke Metals, Inc.

4'-bromo-3-(3,4-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843842
  • Cas Number : 898779-50-5
  • Molecular Weight : 335.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-32
Availability details for
Quantity
  • Availability
  • Add to Cart