RIE156328300 Rieke Metals, Inc.

3'-chloro-3-(3,4-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843844
  • Cas Number : 898779-55-0
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-(3,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-3-4-13(9-12(11)2)5-8-17(20)14-6-7-16(19)15(18)10-14/h3-4,6-7,9-10H,5,8H2,1-2H3
Inchi Key QNYLCVHWQWYTOY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-34
Availability details for
Quantity
  • Availability
  • Add to Cart