RIE156328302 Rieke Metals, Inc.

3-(3,4-dimethylphenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843846
  • Cas Number : 898779-59-4
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-37
Availability details for
Quantity
  • Availability
  • Add to Cart