RIE156328306 Rieke Metals, Inc.

4'-bromo-3-(3,4-dimethylphenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843850
  • Cas Number : 898779-67-4
  • Molecular Weight : 335.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-49
Availability details for
Quantity
  • Availability
  • Add to Cart