RIE156328307 Rieke Metals, Inc.

2'-chloro-3-(3,4-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843851
  • Cas Number : 898779-69-6
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloro-4-fluorophenyl)-3-(3,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-3-4-13(9-12(11)2)5-8-17(20)15-7-6-14(19)10-16(15)18/h3-4,6-7,9-10H,5,8H2,1-2H3
Inchi Key RSYVYOSEMXLFKU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-50
Availability details for
Quantity
  • Availability
  • Add to Cart