RIE156328311 Rieke Metals, Inc.

2',4'-dichloro-3-(3,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843855
  • Cas Number : 898779-77-6
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-dichlorophenyl)-3-(3,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16Cl2O/c1-11-3-4-13(9-12(11)2)5-8-17(20)15-7-6-14(18)10-16(15)19/h3-4,6-7,9-10H,5,8H2,1-2H3
Inchi Key XCLUZWJSUKOTQK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3000-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-54
Availability details for
Quantity
  • Availability
  • Add to Cart