RIE156328312 Rieke Metals, Inc.

2',5'-dichloro-3-(3,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843856
  • Cas Number : 898779-79-8
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-55
Availability details for
Quantity
  • Availability
  • Add to Cart