RIE156328313 Rieke Metals, Inc.

3',4'-dichloro-3-(3,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843857
  • Cas Number : 898779-81-2
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3000-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3000-56
Availability details for
Quantity
  • Availability
  • Add to Cart