RIE156328324 Rieke Metals, Inc.

3-(3,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843867
  • Cas Number : 854692-50-5
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-01
Availability details for
Quantity
  • Availability
  • Add to Cart