RIE156328330 Rieke Metals, Inc.

3-(3,5-dimethylphenyl)-4'-methoxypropiophenone

  • MDL number MFCD03843873
  • Cas Number : 898780-20-6
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3,5-dimethylphenyl)-1-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C18H20O2/c1-13-10-14(2)12-15(11-13)4-9-18(19)16-5-7-17(20-3)8-6-16/h5-8,10-12H,4,9H2,1-3H3
Inchi Key DXWOHFPSJJCHQM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3100-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-07
Availability details for
Quantity
  • Availability
  • Add to Cart