RIE156328337 Rieke Metals, Inc.

3-(3,5-dimethylphenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD03843880
  • Cas Number : 898780-41-1
  • Molecular Weight : 284.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-14
Availability details for
Quantity
  • Availability
  • Add to Cart