RIE156328340 Rieke Metals, Inc.

4'-bromo-3-(3,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843883
  • Cas Number : 898780-50-2
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-19
Availability details for
Quantity
  • Availability
  • Add to Cart