RIE156328342 Rieke Metals, Inc.

4'-chloro-3-(3,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843885
  • Cas Number : 898780-56-8
  • Molecular Weight : 272.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chlorophenyl)-3-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17ClO/c1-12-9-13(2)11-14(10-12)3-8-17(19)15-4-6-16(18)7-5-15/h4-7,9-11H,3,8H2,1-2H3
Inchi Key OVCYONKMUSSWNF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3100-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-21
Availability details for
Quantity
  • Availability
  • Add to Cart