RIE156328354 Rieke Metals, Inc.

2'-chloro-3-(3,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843897
  • Cas Number : 898780-80-8
  • Molecular Weight : 272.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-36
Availability details for
Quantity
  • Availability
  • Add to Cart