RIE156328358 Rieke Metals, Inc.

3-(3,5-dimethylphenyl)-4'-trifluoromethylpropiophenone

  • MDL number MFCD03843901
  • Cas Number : 898780-88-6
  • Molecular Weight : 306.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-45
Availability details for
Quantity
  • Availability
  • Add to Cart