RIE156328359 Rieke Metals, Inc.

4'-bromo-3-(3,5-dimethylphenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843902
  • Cas Number : 898780-90-0
  • Molecular Weight : 335.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-2-fluorophenyl)-3-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16BrFO/c1-11-7-12(2)9-13(8-11)3-6-17(20)15-5-4-14(18)10-16(15)19/h4-5,7-10H,3,6H2,1-2H3
Inchi Key AYKDLSHGZHIDAN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3100-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-49
Availability details for
Quantity
  • Availability
  • Add to Cart