RIE156328361 Rieke Metals, Inc.

3'-chloro-3-(3,5-dimethylphenyl)-5'-fluoropropiophenone

  • MDL number MFCD03843904
  • Cas Number : 898780-94-4
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-51
Availability details for
Quantity
  • Availability
  • Add to Cart