RIE156328362 Rieke Metals, Inc.

4'-chloro-3-(3,5-dimethylphenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843905
  • Cas Number : 898780-96-6
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3100-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-52
Availability details for
Quantity
  • Availability
  • Add to Cart