RIE156328368 Rieke Metals, Inc.

2',4'-difluoro-3-(3,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843911
  • Cas Number : 898781-10-7
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-difluorophenyl)-3-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16F2O/c1-11-7-12(2)9-13(8-11)3-6-17(20)15-5-4-14(18)10-16(15)19/h4-5,7-10H,3,6H2,1-2H3
Inchi Key PVYRHUOOJKEYSB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3100-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3100-58
Availability details for
Quantity
  • Availability
  • Add to Cart