RIE156328373 Rieke Metals, Inc.

cyclopropyl 2-(3,5-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843915
  • Cas Number : 898781-25-4
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropyl-3-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C14H18O/c1-10-7-11(2)9-12(8-10)3-6-14(15)13-4-5-13/h7-9,13H,3-6H2,1-2H3
Inchi Key PPWXFIUFJGYPLY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-3101-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3101-00
Availability details for
Quantity
  • Availability
  • Add to Cart