RIE156328374 Rieke Metals, Inc.

cyclobutyl 2-(3,5-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843916
  • Cas Number : 898781-28-7
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3102-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3102-00
Availability details for
Quantity
  • Availability
  • Add to Cart