RIE156328377 Rieke Metals, Inc.

3-(4-Chloro-3-fluorophenyl)-2',6'-dichloropropiophenone

  • MDL number MFCD08064149
  • Cas Number : 898781-40-3
  • Molecular Weight : 331.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-3300-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-3300-62
Availability details for
Quantity
  • Availability
  • Add to Cart