RIE156328379 Rieke Metals, Inc.

2',6'-dichloro-3-(2-trifluoromethylphenyl)propiophenone

  • MDL number MFCD08064153
  • Cas Number : 898749-86-5
  • Molecular Weight : 347.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-4300-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-4300-62
Availability details for
Quantity
  • Availability
  • Add to Cart