RIE156328382 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)propiophenone

  • MDL number MFCD03843919
  • Cas Number : 898750-01-1
  • Molecular Weight : 262.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-chloro-5-fluorophenyl)-1-phenyl-1-propanone
Inchi Code 1S/C15H12ClFO/c16-13-8-11(9-14(17)10-13)6-7-15(18)12-4-2-1-3-5-12/h1-5,8-10H,6-7H2
Inchi Key QXVGTRSOJXHWSG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-5100-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-01
Availability details for
Quantity
  • Availability
  • Add to Cart