RIE156328384 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-3'-methylpropiophenone

  • MDL number MFCD03843921
  • Cas Number : 898750-07-7
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-chloro-5-fluorophenyl)-1-(3-methylphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO/c1-11-3-2-4-13(7-11)16(19)6-5-12-8-14(17)10-15(18)9-12/h2-4,7-10H,5-6H2,1H3
Inchi Key VUSIZCINELIVHE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-5100-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-03
Availability details for
Quantity
  • Availability
  • Add to Cart