RIE156328386 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-2'-methoxypropiophenone

  • MDL number MFCD03843923
  • Cas Number : 898750-13-5
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-chloro-5-fluorophenyl)-1-(2-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO2/c1-20-16-5-3-2-4-14(16)15(19)7-6-11-8-12(17)10-13(18)9-11/h2-5,8-10H,6-7H2,1H3
Inchi Key BZXQIESLOKSGPK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-5100-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-05
Availability details for
Quantity
  • Availability
  • Add to Cart