RIE156328387 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-3'-methoxypropiophenone

  • MDL number MFCD03843924
  • Cas Number : 898750-16-8
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-chloro-5-fluorophenyl)-1-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO2/c1-20-15-4-2-3-12(9-15)16(19)6-5-11-7-13(17)10-14(18)8-11/h2-4,7-10H,5-6H2,1H3
Inchi Key LSTJAQXGKCPFLQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-5100-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-06
Availability details for
Quantity
  • Availability
  • Add to Cart