RIE156328388 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-4'-methoxypropiophenone

  • MDL number MFCD03843925
  • Cas Number : 898750-19-1
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-07
Availability details for
Quantity
  • Availability
  • Add to Cart