RIE156328389 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-2'-cyanopropiophenone

  • MDL number MFCD03843926
  • Cas Number : 898750-22-6
  • Molecular Weight : 287.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-08
Availability details for
Quantity
  • Availability
  • Add to Cart