RIE156328390 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-3'-cyanopropiophenone

  • MDL number MFCD03843927
  • Cas Number : 898750-25-9
  • Molecular Weight : 287.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-09
Availability details for
Quantity
  • Availability
  • Add to Cart