RIE156328402 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843939
  • Cas Number : 898750-61-3
  • Molecular Weight : 280.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-23
Availability details for
Quantity
  • Availability
  • Add to Cart