RIE156328404 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-2',4'-dimethylpropiophenone

  • MDL number MFCD03843941
  • Cas Number : 898750-67-9
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-27
Availability details for
Quantity
  • Availability
  • Add to Cart