RIE156328408 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-3',5'-dimethylpropiophenone

  • MDL number MFCD03843945
  • Cas Number : 898750-79-3
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-31
Availability details for
Quantity
  • Availability
  • Add to Cart