RIE156328409 Rieke Metals, Inc.

4'-bromo-3-(3-chloro-5-fluorophenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843946
  • Cas Number : 898750-82-8
  • Molecular Weight : 359.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-3-fluorophenyl)-3-(3-chloro-5-fluorophenyl)-1-propanone
Inchi Code 1S/C15H10BrClF2O/c16-13-3-2-10(7-14(13)19)15(20)4-1-9-5-11(17)8-12(18)6-9/h2-3,5-8H,1,4H2
Inchi Key TWCKHWLVVDGDHV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-5100-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-32
Availability details for
Quantity
  • Availability
  • Add to Cart