RIE156328415 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD03843952
  • Cas Number : 898751-00-3
  • Molecular Weight : 330.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-44
Availability details for
Quantity
  • Availability
  • Add to Cart