RIE156328417 Rieke Metals, Inc.

4'-bromo-3-(3-chloro-5-fluorophenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843954
  • Cas Number : 898751-08-1
  • Molecular Weight : 359.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-49
Availability details for
Quantity
  • Availability
  • Add to Cart